The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024963.5 | NP_079239.3 | 4 - 5 | 2125 | 85 |
GenBank mRNA | BC004228.1 | - | 2 - 3 | 578 | 85 |
BC013435.1 | - | 2 - 3 | 727 | 85 |