The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006503.3 | NP_006494.1 | 3 - 4 | 523 | 67 |
NM_153001.2 | NP_694546.1 | 3 - 4 | 430 | 67 | |
GenBank mRNA | AF038965.1 | - | 3 - 4 | 347 | 67 |
AK290285.1 | - | 3 - 4 | 331 | 67 | |
AK310135.1 | - | 3 - 4 | 348 | 67 | |
AK313183.1 | - | 3 - 4 | 365 | 67 | |
BC000343.2 | - | 3 - 4 | 345 | 67 | |
BC010396.1 | - | 3 - 4 | 269 | 67 | |
BC014488.1 | - | 3 - 4 | 345 | 67 | |
BC018811.2 | - | 3 - 4 | 345 | 67 | |
BI828251.1 | - | 3 - 4 | 523 | 67 | |
BT007232.1 | - | 3 - 4 | 325 | 67 | |
U27515.1 | - | 3 - 4 | 337 | 67 |