The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006503.3 | NP_006494.1 | 4 - 5 | 671 | 77 |
NM_153001.2 | NP_694546.1 | 4 - 5 | 578 | 77 | |
GenBank mRNA | AF038965.1 | - | 4 - 5 | 495 | 77 |
AK290285.1 | - | 4 - 5 | 479 | 77 | |
AK310135.1 | - | 4 - 5 | 77 | ||
AK313183.1 | - | 4 - 5 | 513 | 77 | |
BC000343.2 | - | 4 - 5 | 493 | 77 | |
BC010396.1 | - | 4 - 5 | 417 | 77 | |
BC014488.1 | - | 4 - 5 | 493 | 77 | |
BC018811.2 | - | 4 - 5 | 493 | 77 | |
BI828251.1 | - | 4 - 5 | 671 | 77 | |
BT007232.1 | - | 4 - 5 | 473 | 77 | |
U27515.1 | - | 4 - 5 | 485 | 77 |