Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2F1
Entrez Gene ID
1572
Gene Name
cytochrome P450 family 2 subfamily F member 1
Gene Aliases
C2F1, CYP2F, CYPIIF1
Chromosome Location
Chr.19: 41114292 - 41128381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000774.4 NP_000765.2 2 - 3 245 63
NR_135528.1 2 - 3 245 63
XM_011526551.2 XP_011524853.1 2 - 3 385 63
XM_011526552.2 XP_011524854.1 2 - 3 388 63
XM_011526553.2 XP_011524855.1 1 - 2 576 63
XM_011526555.2 XP_011524857.1 1 - 2 576 63
XM_017026384.1 XP_016881873.1 1 - 2 576 63
XM_017026385.1 XP_016881874.1 1 - 2 576 63
XM_017026386.1 XP_016881875.1 1 - 2 576 63
XM_017026387.1 XP_016881876.1 1 - 2 576 63
GenBank mRNA AK298082.1 - 2 - 3 248 63
AK316498.1 - 2 - 3 245 63
BC109056.1 - 2 - 3 222 63
DC413790.1 - 2 - 3 245 63
J02906.1 - 2 - 3 228 63

View Full Product Details