The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166159.1 | NP_001159631.1 | 4 - 5 | 751 | 90 |
NM_004550.4 | NP_004541.1 | 5 - 6 | 960 | 90 | |
XM_005245208.2 | XP_005245265.1 | 5 - 6 | 862 | 90 | |
XM_005245209.1 | XP_005245266.1 | 3 - 4 | 431 | 90 | |
XM_017001357.1 | XP_016856846.1 | 4 - 5 | 1226 | 90 | |
GenBank mRNA | AF013160.1 | - | 4 - 5 | 518 | 90 |
AF050640.1 | - | 4 - 5 | 518 | 90 | |
AK222618.1 | - | 4 - 5 | 517 | 90 | |
AK301641.1 | - | 4 - 5 | 565 | 90 | |
AK314807.1 | - | 4 - 5 | 524 | 90 | |
AK315977.1 | - | 3 - 4 | 419 | 90 | |
BC000170.2 | - | 4 - 5 | 519 | 90 | |
BC001456.1 | - | 4 - 5 | 513 | 90 | |
BC008868.2 | - | 5 - 6 | 960 | 90 |