The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122636.1 | NP_001116108.1 | 4 - 5 | 874 | 57 |
XM_017019371.1 | XP_016874860.1 | 3 - 4 | 606 | 57 | |
XM_017019372.1 | XP_016874861.1 | 3 - 4 | 566 | 57 | |
XM_017019373.1 | XP_016874862.1 | 4 - 5 | 923 | 57 | |
XM_017019374.1 | XP_016874863.1 | 3 - 4 | 543 | 57 | |
XM_017019375.1 | XP_016874864.1 | 4 - 5 | 877 | 57 | |
XM_017019376.1 | XP_016874865.1 | 4 - 5 | 877 | 57 | |
GenBank mRNA | AK055773.1 | - | 3 - 4 | 531 | 57 |
BC157845.1 | - | 4 - 5 | 904 | 57 | |
BC171844.1 | - | 4 - 5 | 904 | 57 | |
BM925654.1 | - | 4 - 5 | 652 | 57 |