The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282982.1 | NP_001269911.1 | 10 - 11 | 923 | 99 |
NM_020908.2 | NP_065959.1 | 12 - 13 | 2141 | 99 | |
XM_005254625.3 | XP_005254682.1 | 12 - 13 | 2189 | 99 | |
XM_005254628.3 | XP_005254685.1 | 12 - 13 | 2189 | 99 | |
XM_005254633.2 | XP_005254690.1 | 12 - 13 | 2159 | 99 | |
GenBank mRNA | AB046767.1 | - | 12 - 13 | 2123 | 99 |
AK096779.1 | - | 10 - 11 | 923 | 99 |