Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MTSS1
- Entrez Gene ID
-
9788
- Gene Name
- metastasis suppressor 1
- Gene Aliases
-
MIM, MIMA, MIMB
- Chromosome Location
-
Chr.8: 124550770 - 124728507 on Build GRCh38
View Full Product Details