The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282971.1 | NP_001269900.1 | 11 - 12 | 1597 | 98 |
NM_014751.5 | NP_055566.3 | 10 - 11 | 1585 | 98 | |
XM_005251111.1 | XP_005251168.1 | 11 - 12 | 1563 | 98 | |
XM_006716700.1 | XP_006716763.1 | 11 - 12 | 1560 | 98 | |
XM_006716701.1 | XP_006716764.1 | 10 - 11 | 1551 | 98 | |
XM_006716702.1 | XP_006716765.1 | 11 - 12 | 1563 | 98 | |
XM_006716704.1 | XP_006716767.1 | 11 - 12 | 1560 | 98 | |
XM_006716705.1 | XP_006716768.1 | 11 - 12 | 1563 | 98 | |
XM_011517403.1 | XP_011515705.1 | 10 - 11 | 1017 | 98 | |
XM_011517404.2 | XP_011515706.1 | 9 - 10 | 12729 | 98 | |
XM_011517407.1 | XP_011515709.1 | 2 - 3 | 268 | 98 | |
XM_017014086.1 | XP_016869575.1 | 10 - 11 | 1548 | 98 | |
XM_017014090.1 | XP_016869579.1 | 9 - 10 | 2109 | 98 | |
XM_017014091.1 | XP_016869580.1 | 11 - 12 | 997 | 98 | |
XM_017014093.1 | XP_016869582.1 | 10 - 11 | 1548 | 98 | |
XM_017014094.1 | XP_016869583.1 | 10 - 11 | 1551 | 98 | |
GenBank mRNA | AF086645.1 | - | 10 - 11 | 1496 | 98 |
AF116674.1 | - | 4 - 5 | 542 | 98 | |
AK027015.1 | - | 11 - 12 | 1363 | 98 | |
AK302036.1 | - | 4 - 5 | 841 | 98 | |
AK310792.1 | - | 2 - 3 | 307 | 98 |