The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000395.2 | NP_000386.1 | 12 - 13 | 1677 | 81 |
XM_005261340.3 | XP_005261397.1 | 12 - 13 | 1754 | 81 | |
XM_011529903.2 | XP_011528205.1 | 12 - 13 | 1790 | 81 | |
XM_011529904.2 | XP_011528206.1 | 12 - 13 | 1772 | 81 | |
GenBank mRNA | AK302993.1 | - | 10 - 11 | 1339 | 81 |
BC070085.1 | - | 12 - 13 | 1677 | 81 | |
CR456428.1 | - | 11 - 12 | 1551 | 81 | |
M59941.1 | - | 11 - 12 | 1488 | 81 |