The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000395.2 | NP_000386.1 | 7 - 8 | 1071 | 123 |
XM_011529904.2 | XP_011528206.1 | 7 - 8 | 1130 | 123 | |
GenBank mRNA | BC070085.1 | - | 7 - 8 | 1071 | 123 |
M59941.1 | - | 6 - 7 | 882 | 123 |