The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002736.2 | NP_002727.2 | 6 - 7 | 921 | 142 |
XM_011516398.1 | XP_011514700.1 | 5 - 6 | 842 | 142 | |
GenBank mRNA | AK128639.1 | - | 7 - 8 | 731 | 142 |
AK291441.1 | - | 6 - 7 | 772 | 142 | |
BC075800.1 | - | 6 - 7 | 960 | 142 | |
M31158.1 | - | 6 - 7 | 912 | 142 |