The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002736.2 | NP_002727.2 | 8 - 9 | 1092 | 152 |
XM_011516398.1 | XP_011514700.1 | 7 - 8 | 1013 | 152 | |
GenBank mRNA | AK291441.1 | - | 8 - 9 | 943 | 152 |
BC075800.1 | - | 8 - 9 | 1131 | 152 |