The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007235.4 | NP_009166.2 | 3 - 4 | 643 | 217 |
XM_017018748.1 | XP_016874237.1 | 3 - 4 | 503 | 217 | |
GenBank mRNA | AF039022.1 | - | 2 - 3 | 137 | 217 |
AK292884.1 | - | 3 - 4 | 456 | 217 | |
AK308108.1 | - | 3 - 4 | 464 | 147 | |
BC020569.1 | - | 2 - 3 | 222 | 217 | |
CR979468.1 | - | 3 - 4 | 643 | 217 | |
Y16414.1 | - | 2 - 3 | 195 | 217 |