The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007235.4 | NP_009166.2 | 5 - 6 | 777 | 96 |
XM_017018748.1 | XP_016874237.1 | 5 - 6 | 637 | 96 | |
GenBank mRNA | AF039022.1 | - | 4 - 5 | 271 | 96 |
AK292884.1 | - | 5 - 6 | 590 | 96 | |
BC020569.1 | - | 4 - 5 | 356 | 96 | |
Y16414.1 | - | 4 - 5 | 329 | 96 |