The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202498.1 | NP_001189427.1 | 3 - 4 | 219 | 114 |
NM_003344.3 | NP_003335.1 | 3 - 4 | 615 | 114 | |
GenBank mRNA | AB451490.1 | - | 3 - 4 | 210 | 114 |
AK316590.1 | - | 3 - 4 | 198 | 114 | |
BC006277.2 | - | 3 - 4 | 527 | 114 | |
BI088146.1 | - | 3 - 4 | 219 | 114 | |
BT006756.1 | - | 3 - 4 | 210 | 114 | |
KJ534959.1 | - | 3 - 4 | 210 | 114 | |
Z29328.1 | - | 3 - 4 | 263 | 114 | |
Z29330.1 | - | 3 - 4 | 263 | 114 |