Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11orf80
Entrez Gene ID
79703
Gene Name
chromosome 11 open reading frame 80
Gene Aliases
TOP6BL, TOPOVIBL
Chromosome Location
Chr.11: 66744736 - 66843516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK026184.1 - 11 - 12 1267 74
AK225435.1 - 11 - 12 1267 74

View Full Product Details