Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY3
Entrez Gene ID
23001
Gene Name
WD repeat and FYVE domain containing 3
Gene Aliases
ALFY, ZFYVE25
Chromosome Location
Chr.4: 84669537 - 84966391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014991.4 NP_055806.2 22 - 23 4077 125
XM_005262858.4 XP_005262915.1 24 - 25 4118 125
XM_011531757.2 XP_011530059.1 23 - 24 4029 125
XM_011531759.2 XP_011530061.1 22 - 23 3991 125
XM_011531760.2 XP_011530062.1 22 - 23 3929 125
XM_011531761.2 XP_011530063.1 23 - 24 4029 125
XM_011531762.2 XP_011530064.1 23 - 24 4029 125
XM_011531763.2 XP_011530065.1 21 - 22 3875 125
XM_011531764.2 XP_011530066.1 22 - 23 3945 125
XM_011531765.2 XP_011530067.1 20 - 21 3731 125
XM_011531766.2 XP_011530068.1 23 - 24 4029 125
XM_011531767.2 XP_011530069.1 23 - 24 4029 125
XM_017007906.1 XP_016863395.1 25 - 26 4246 125
XM_017007907.1 XP_016863396.1 25 - 26 4243 125
XM_017007908.1 XP_016863397.1 24 - 25 4175 125
XM_017007909.1 XP_016863398.1 10 - 11 1871 125
GenBank mRNA AF538685.1 - 21 - 22 3969 125

View Full Product Details