The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014991.4 | NP_055806.2 | 24 - 25 | 4475 | 90 |
XM_005262858.4 | XP_005262915.1 | 26 - 27 | 4516 | 90 | |
XM_011531757.2 | XP_011530059.1 | 25 - 26 | 4427 | 90 | |
XM_011531759.2 | XP_011530061.1 | 24 - 25 | 4389 | 90 | |
XM_011531760.2 | XP_011530062.1 | 24 - 25 | 4327 | 90 | |
XM_011531761.2 | XP_011530063.1 | 25 - 26 | 4427 | 90 | |
XM_011531762.2 | XP_011530064.1 | 25 - 26 | 4427 | 90 | |
XM_011531763.2 | XP_011530065.1 | 23 - 24 | 4273 | 90 | |
XM_011531764.2 | XP_011530066.1 | 24 - 25 | 4343 | 90 | |
XM_011531765.2 | XP_011530067.1 | 22 - 23 | 4129 | 90 | |
XM_011531766.2 | XP_011530068.1 | 25 - 26 | 4427 | 90 | |
XM_011531767.2 | XP_011530069.1 | 25 - 26 | 4427 | 90 | |
XM_017007906.1 | XP_016863395.1 | 27 - 28 | 4644 | 90 | |
XM_017007907.1 | XP_016863396.1 | 27 - 28 | 4641 | 90 | |
XM_017007908.1 | XP_016863397.1 | 26 - 27 | 4573 | 90 | |
XM_017007909.1 | XP_016863398.1 | 12 - 13 | 2269 | 90 | |
GenBank mRNA | AF538685.1 | - | 23 - 24 | 4367 | 90 |