The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014991.4 | NP_055806.2 | 5 - 6 | 710 | 64 |
XM_011531763.2 | XP_011530065.1 | 4 - 5 | 508 | 64 | |
XM_011531765.2 | XP_011530067.1 | 3 - 4 | 364 | 64 | |
GenBank mRNA | AF538685.1 | - | 4 - 5 | 602 | 64 |
AK092019.1 | - | 3 - 4 | 364 | 64 |