The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005689.2 | NP_005680.1 | 6 - 7 | 70 | |
GenBank mRNA | AF070598.1 | - | 2 - 3 | 70 | |
AF308472.1 | - | 6 - 7 | 70 | ||
AJ289233.2 | - | 6 - 7 | 70 | ||
AK026067.1 | - | 3 - 4 | 409 | 70 | |
AK057026.1 | - | 6 - 7 | 70 | ||
AK172812.1 | - | 6 - 7 | 70 | ||
AK303230.1 | - | 5 - 6 | 70 | ||
BC000559.2 | - | 6 - 7 | 70 |