Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDF2L1
Entrez Gene ID
23753
Gene Name
stromal cell derived factor 2 like 1
Gene Aliases
-
Chromosome Location
Chr.22: 21642253 - 21644299 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022044.2 NP_071327.2 1 - 2 273 55
GenBank mRNA AB043007.1 - 1 - 2 209 55
AF277316.1 - 1 - 2 197 55
AY359118.1 - 1 - 2 240 55
BC132849.1 - 1 - 2 265 55
BC132851.1 - 1 - 2 265 55
CR456570.1 - 1 - 2 198 55

View Full Product Details