Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
Chromosome Location
Chr.12: 42081845 - 42144871 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099650.1 NP_001093120.1 2 - 3 620 139
NM_173601.1 NP_775872.1 3 - 4 713 139
XM_017019211.1 XP_016874700.1 3 - 4 452 139
GenBank mRNA AK125141.1 - 2 - 3 473 139
BC030023.1 - 3 - 4 721 139
BC039145.1 - 2 - 3 460 139

View Full Product Details