Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CASQ1
Entrez Gene ID
844
Gene Name
calsequestrin 1
Gene Aliases
CASQ, PDIB1, VMCQA
Chromosome Location
Chr.1: 160190495 - 160201886 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001231.4 NP_001222.3 4 - 5 839 62
GenBank mRNA AK300713.1 - 3 - 4 643 62
AK313250.1 - 4 - 5 742 62
BC022289.1 - 4 - 5 725 62
S73775.1 - 4 - 5 729 62

View Full Product Details