The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042717.2 | NP_001036182.1 | 3 - 4 | 376 | 149 |
NM_033107.3 | NP_149098.2 | 5 - 6 | 613 | 149 | |
GenBank mRNA | AF351613.1 | - | 3 - 4 | 377 | 149 |
AK095561.1 | - | 5 - 6 | 603 | 149 | |
AK294325.1 | - | 3 - 4 | 326 | 149 | |
AL525056.3 | - | 5 - 6 | 555 | 149 | |
BC004923.2 | - | 4 - 5 | 510 | 149 | |
BC021573.1 | - | 4 - 5 | 510 | 149 | |
BC107714.1 | - | 5 - 6 | 597 | 149 | |
BC144162.1 | - | 3 - 4 | 323 | 149 | |
CR933597.1 | - | 4 - 5 | 427 | 149 |