The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000210.3 | NP_000201.2 | 16 - 17 | 2477 | 145 |
NM_001079818.2 | NP_001073286.1 | 16 - 17 | 2477 | 145 | |
NM_001316306.1 | NP_001303235.1 | 16 - 17 | 2163 | 145 | |
XM_006712510.1 | XP_006712573.1 | 16 - 17 | 2446 | 145 | |
XM_006712511.1 | XP_006712574.1 | 16 - 17 | 2446 | 145 | |
XM_017004005.1 | XP_016859494.1 | 16 - 17 | 2173 | 145 | |
XM_017004006.1 | XP_016859495.1 | 16 - 17 | 2173 | 145 | |
XM_017004007.1 | XP_016859496.1 | 16 - 17 | 2173 | 145 | |
XM_017004008.1 | XP_016859497.1 | 16 - 17 | 2173 | 145 | |
GenBank mRNA | AB208842.1 | - | 16 - 17 | 2144 | 145 |
AK294436.1 | - | 12 - 13 | 1565 | 145 | |
AK296496.1 | - | 16 - 17 | 2099 | 145 | |
BC050585.1 | - | 10 - 11 | 1128 | 145 | |
BC136455.1 | - | 16 - 17 | 2351 | 145 | |
BC136456.1 | - | 16 - 17 | 2351 | 145 | |
X53586.1 | - | 16 - 17 | 2389 | 145 | |
X59512.1 | - | 16 - 17 | 145 |