Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITGA6
Entrez Gene ID
3655
Gene Name
integrin subunit alpha 6
Gene Aliases
CD49f, ITGA6B, VLA-6
Chromosome Location
Chr.2: 172427354 - 172506455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000210.3 NP_000201.2 20 - 21 2909 148
NM_001079818.2 NP_001073286.1 20 - 21 2909 148
NM_001316306.1 NP_001303235.1 20 - 21 2595 148
XM_017004005.1 XP_016859494.1 20 - 21 2605 148
XM_017004007.1 XP_016859496.1 20 - 21 2605 148
GenBank mRNA AB208842.1 - 20 - 21 2576 148
AK294436.1 - 16 - 17 1997 148
AK296496.1 - 20 - 21 2531 148
BC136455.1 - 20 - 21 2783 148
BC136456.1 - 20 - 21 2783 148
X53586.1 - 20 - 21 2821 148
X59512.1 - 20 - 21 2657 148

View Full Product Details