The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000210.3 | NP_000201.2 | 3 - 4 | 615 | 81 |
NM_001079818.2 | NP_001073286.1 | 3 - 4 | 615 | 81 | |
NM_001316306.1 | NP_001303235.1 | 3 - 4 | 316 | 81 | |
XM_006712510.1 | XP_006712573.1 | 3 - 4 | 584 | 81 | |
XM_006712511.1 | XP_006712574.1 | 3 - 4 | 584 | 81 | |
XM_017004005.1 | XP_016859494.1 | 3 - 4 | 311 | 81 | |
XM_017004006.1 | XP_016859495.1 | 3 - 4 | 311 | 81 | |
XM_017004007.1 | XP_016859496.1 | 3 - 4 | 311 | 81 | |
XM_017004008.1 | XP_016859497.1 | 3 - 4 | 311 | 81 | |
GenBank mRNA | AB208842.1 | - | 3 - 4 | 283 | 81 |
AK296496.1 | - | 3 - 4 | 252 | 81 | |
AK311695.1 | - | 3 - 4 | 316 | 81 | |
BC007002.1 | - | 3 - 4 | 581 | 81 | |
BC136455.1 | - | 3 - 4 | 489 | 81 | |
BC136456.1 | - | 3 - 4 | 489 | 81 | |
X53586.1 | - | 3 - 4 | 527 | 81 | |
X59512.1 | - | 3 - 4 | 363 | 81 |