Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC31A
Entrez Gene ID
22872
Gene Name
SEC31 homolog A, COPII coat complex component
Gene Aliases
ABP125, ABP130, HSPC275, HSPC334, SEC31L1
Chromosome Location
Chr.4: 82818509 - 82900571 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077206.3 NP_001070674.1 6 - 7 840 86
NM_001077207.3 NP_001070675.1 6 - 7 840 86
NM_001077208.3 NP_001070676.1 6 - 7 840 86
NM_001191049.2 NP_001177978.1 5 - 6 757 86
NM_001300744.2 NP_001287673.1 6 - 7 840 86
NM_001300745.2 NP_001287674.1 6 - 7 840 86
NM_001318119.1 NP_001305048.1 6 - 7 837 86
NM_001318120.1 NP_001305049.1 8 - 9 1054 86
NM_016211.4 NP_057295.2 6 - 7 840 86
GenBank mRNA AB018358.1 - 6 - 7 696 86
AB018359.1 - 6 - 7 696 86
AB020712.1 - 8 - 9 1021 86
AF139184.1 - 5 - 6 643 86
AK002161.1 - 6 - 7 713 86
AK225934.1 - 6 - 7 700 86
AK295810.1 - 5 - 6 655 86
BC047883.2 - 6 - 7 744 86
BC084583.1 - 6 - 7 679 86
BC117221.1 - 6 - 7 708 86
BC143489.1 - 6 - 7 705 86
BC143491.1 - 6 - 7 708 86
BC143492.2 - 6 - 7 708 86
CA397330.1 - 2 - 3 187 86
CR933696.1 - 6 - 7 713 86

View Full Product Details