Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYCP3
Entrez Gene ID
50511
Gene Name
synaptonemal complex protein 3
Gene Aliases
COR1, RPRGL4, SCP3, SPGF4
Chromosome Location
Chr.12: 101728648 - 101739722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177948.1 NP_001171419.1 7 - 8 680 139
NM_001177949.1 NP_001171420.1 7 - 8 695 139
NM_153694.4 NP_710161.1 7 - 8 691 139
XM_005268922.4 XP_005268979.3 7 - 8 941 139
XM_005268924.1 XP_005268981.1 7 - 8 679 139
XM_005268925.1 XP_005268982.1 7 - 8 684 139
XM_005268926.3 XP_005268983.1 7 - 8 663 139
XM_005268927.2 XP_005268984.1 7 - 8 652 139
XM_011538421.2 XP_011536723.2 7 - 8 863 139
XM_017019368.1 XP_016874857.1 7 - 8 938 139
GenBank mRNA AF492003.1 - 7 - 8 636 139
AF517774.1 - 7 - 8 634 139
AK125930.1 - 4 - 5 3253 139
BC062662.1 - 7 - 8 686 139

View Full Product Details