The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000437.3 | NP_000428.2 | 2 - 3 | 268 | 120 |
XM_006710670.3 | XP_006710733.1 | 2 - 3 | 333 | 120 | |
XM_011541528.2 | XP_011539830.1 | 3 - 4 | 399 | 120 | |
XM_017001390.1 | XP_016856879.1 | 3 - 4 | 391 | 120 | |
XM_017001391.1 | XP_016856880.1 | 2 - 3 | 166 | 120 | |
GenBank mRNA | AK313815.1 | - | 2 - 3 | 195 | 120 |
BC001158.1 | - | 2 - 3 | 213 | 120 | |
D87845.1 | - | 2 - 3 | 291 | 120 | |
U89386.1 | - | 2 - 3 | 167 | 120 |