The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000437.3 | NP_000428.2 | 4 - 5 | 517 | 76 |
XM_006710670.3 | XP_006710733.1 | 4 - 5 | 582 | 76 | |
XM_011541528.2 | XP_011539830.1 | 5 - 6 | 648 | 76 | |
GenBank mRNA | AK313815.1 | - | 4 - 5 | 444 | 76 |
BC001158.1 | - | 4 - 5 | 462 | 76 | |
D87845.1 | - | 4 - 5 | 540 | 76 | |
U89386.1 | - | 4 - 5 | 416 | 76 |