The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000613.2 | NP_000604.1 | 5 - 6 | 555 | 59 |
GenBank mRNA | AK313648.1 | - | 5 - 6 | 518 | 59 |
BC005395.1 | - | 5 - 6 | 555 | 59 | |
J03048.1 | - | 5 - 6 | 491 | 59 | |
X02537.1 | - | 5 - 6 | 431 | 59 |