Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Chromosome Location
Chr.1: 11106531 - 11262551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004958.3 NP_004949.1 45 - 46 6472 83
XM_005263438.2 XP_005263495.1 45 - 46 6479 83
XM_017000900.1 XP_016856389.1 42 - 43 5935 83
XM_017000901.1 XP_016856390.1 43 - 44 6223 83
GenBank mRNA AB209995.1 - 45 - 46 6453 83
AK126762.1 - 7 - 8 1768 83
BC117166.1 - 44 - 45 6369 83
L34075.1 - 45 - 46 6430 83
L35478.1 - 3 - 4 393 83
U88966.1 - 45 - 46 6427 83

View Full Product Details