Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Chromosome Location
Chr.1: 11106531 - 11262551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004958.3 NP_004949.1 46 - 47 6647 78
XM_005263438.2 XP_005263495.1 46 - 47 6654 78
XM_017000900.1 XP_016856389.1 43 - 44 6110 78
XM_017000901.1 XP_016856390.1 44 - 45 6398 78
GenBank mRNA AB209995.1 - 46 - 47 6628 78
AK126762.1 - 8 - 9 1943 78
BC117166.1 - 45 - 46 6544 78
L34075.1 - 46 - 47 6605 78
U88966.1 - 46 - 47 6602 78

View Full Product Details