The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 52 - 53 | 7287 | 105 |
XM_005263438.2 | XP_005263495.1 | 52 - 53 | 7294 | 105 | |
XM_017000900.1 | XP_016856389.1 | 49 - 50 | 6750 | 105 | |
XM_017000901.1 | XP_016856390.1 | 50 - 51 | 7038 | 105 | |
GenBank mRNA | AB209995.1 | - | 52 - 53 | 7268 | 105 |
AK126762.1 | - | 14 - 15 | 2583 | 105 | |
BC117166.1 | - | 51 - 52 | 7184 | 105 | |
L34075.1 | - | 52 - 53 | 7245 | 105 | |
U88966.1 | - | 52 - 53 | 7242 | 105 |