Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Chromosome Location
Chr.1: 11106531 - 11262551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004958.3 NP_004949.1 7 - 8 1239 74
XM_005263438.2 XP_005263495.1 7 - 8 1246 74
XM_011541166.2 XP_011539468.1 7 - 8 1239 74
XM_017000900.1 XP_016856389.1 4 - 5 702 74
XM_017000902.1 XP_016856391.1 7 - 8 1239 74
GenBank mRNA AB209995.1 - 7 - 8 1220 74
AK302863.1 - 7 - 8 1231 74
AK304273.1 - 7 - 8 1228 74
BC117166.1 - 6 - 7 1136 74
L34075.1 - 7 - 8 1197 74

View Full Product Details