Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTOR
Entrez Gene ID
2475
Gene Name
mechanistic target of rapamycin
Gene Aliases
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Chromosome Location
Chr.1: 11106531 - 11262551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004958.3 NP_004949.1 9 - 10 1537 103
XM_005263438.2 XP_005263495.1 9 - 10 1544 103
XM_011541166.2 XP_011539468.1 9 - 10 1537 103
XM_017000900.1 XP_016856389.1 6 - 7 1000 103
XM_017000901.1 XP_016856390.1 7 - 8 1288 103
XM_017000902.1 XP_016856391.1 9 - 10 1537 103
GenBank mRNA AB209995.1 - 9 - 10 1518 103
AK302863.1 - 9 - 10 1529 103
AK304273.1 - 9 - 10 1526 103
BC117166.1 - 8 - 9 1434 103
L34075.1 - 9 - 10 1495 103

View Full Product Details