The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143821.2 | NP_001137293.2 | 11 - 12 | 1866 | 74 |
NM_001256470.1 | NP_001243399.1 | 12 - 13 | 1884 | 74 | |
NM_001256787.1 | NP_001243716.1 | 9 - 10 | 1650 | 74 | |
NM_019012.5 | NP_061885.2 | 11 - 12 | 1866 | 74 | |
XM_005253400.1 | XP_005253457.1 | 11 - 12 | 1826 | 74 | |
XM_006719091.1 | XP_006719154.1 | 11 - 12 | 1826 | 74 | |
XM_006719096.1 | XP_006719159.1 | 11 - 12 | 1826 | 74 | |
XM_011520714.1 | XP_011519016.1 | 11 - 12 | 1826 | 74 | |
XM_011520715.1 | XP_011519017.1 | 11 - 12 | 1826 | 74 | |
XM_011520716.1 | XP_011519018.1 | 11 - 12 | 1826 | 74 | |
XM_011520717.1 | XP_011519019.1 | 11 - 12 | 1826 | 74 | |
XM_017019495.1 | XP_016874984.1 | 11 - 12 | 1826 | 74 | |
XM_017019496.1 | XP_016874985.1 | 11 - 12 | 1826 | 74 | |
XM_017019497.1 | XP_016874986.1 | 9 - 10 | 1640 | 74 | |
XM_017019498.1 | XP_016874987.1 | 7 - 8 | 1443 | 74 | |
XM_017019499.1 | XP_016874988.1 | 11 - 12 | 1826 | 74 | |
XM_017019500.1 | XP_016874989.1 | 11 - 12 | 1826 | 74 | |
XM_017019501.1 | XP_016874990.1 | 11 - 12 | 1826 | 74 | |
GenBank mRNA | AB051473.2 | - | 11 - 12 | 1762 | 74 |
AB642244.1 | - | 12 - 13 | 1850 | 74 | |
AF302150.1 | - | 11 - 12 | 1764 | 74 | |
AK056054.1 | - | 2 - 3 | 518 | 74 | |
AK294739.1 | - | 9 - 10 | 1651 | 74 | |
AK295151.1 | - | 12 - 13 | 1808 | 74 | |
AK296277.1 | - | 9 - 10 | 1643 | 74 | |
AK297391.1 | - | 9 - 10 | 1507 | 74 | |
BC000969.2 | - | 6 - 7 | 1285 | 74 | |
BC004164.2 | - | 2 - 3 | 551 | 74 | |
BC127091.1 | - | 10 - 11 | 1691 | 74 | |
BC127092.1 | - | 11 - 12 | 1764 | 74 |