Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA5
Entrez Gene ID
54477
Gene Name
pleckstrin homology domain containing A5
Gene Aliases
PEPP-2, PEPP2
Chromosome Location
Chr.12: 19129680 - 19376400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143821.2 NP_001137293.2 19 - 20 2637 151
NM_019012.5 NP_061885.2 17 - 18 2463 151
XM_011520716.1 XP_011519018.1 21 - 22 2714 151
XM_011520717.1 XP_011519019.1 19 - 20 2597 151
XM_017019498.1 XP_016874987.1 16 - 17 2295 151
GenBank mRNA AB051473.2 - 19 - 20 2533 151
AF302150.1 - 17 - 18 2361 151
AK297391.1 - 15 - 16 2104 151
BC004164.2 - 8 - 9 1148 151
BC013133.2 - 6 - 7 595 151
BC127091.1 - 16 - 17 2288 151
BC127092.1 - 17 - 18 2361 151

View Full Product Details