Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHA5
Entrez Gene ID
54477
Gene Name
pleckstrin homology domain containing A5
Gene Aliases
PEPP-2, PEPP2
Chromosome Location
Chr.12: 19129680 - 19376400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143821.2 NP_001137293.2 4 - 5 422 84
NM_001256470.1 NP_001243399.1 4 - 5 422 84
NM_001256787.1 NP_001243716.1 2 - 3 206 84
NM_019012.5 NP_061885.2 4 - 5 422 84
XM_005253400.1 XP_005253457.1 4 - 5 382 84
XM_006719091.1 XP_006719154.1 4 - 5 382 84
XM_006719093.1 XP_006719156.1 4 - 5 382 84
XM_006719096.1 XP_006719159.1 4 - 5 382 84
XM_011520714.1 XP_011519016.1 4 - 5 382 84
XM_011520715.1 XP_011519017.1 4 - 5 382 84
XM_011520716.1 XP_011519018.1 4 - 5 382 84
XM_011520717.1 XP_011519019.1 4 - 5 382 84
XM_017019495.1 XP_016874984.1 4 - 5 382 84
XM_017019496.1 XP_016874985.1 4 - 5 382 84
XM_017019497.1 XP_016874986.1 2 - 3 196 84
XM_017019499.1 XP_016874988.1 4 - 5 382 84
XM_017019500.1 XP_016874989.1 4 - 5 382 84
XM_017019501.1 XP_016874990.1 4 - 5 382 84
GenBank mRNA AB051473.2 - 4 - 5 318 84
AB642244.1 - 4 - 5 388 84
AF302150.1 - 4 - 5 320 84
AK294739.1 - 2 - 3 207 84
AK295151.1 - 4 - 5 346 84
AK296277.1 - 2 - 3 199 84
BC070174.1 - 4 - 5 365 84
BC127091.1 - 4 - 5 320 84
BC127092.1 - 4 - 5 320 84
DC311342.1 - 2 - 3 167 84
DR000821.1 - 4 - 5 407 84

View Full Product Details