Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
Chromosome Location
Chr.11: 6621324 - 6655849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003737.3 NP_003728.1 20 - 21 7694 61
GenBank mRNA AB053446.1 - 20 - 21 7697 61
AL831828.1 - 7 - 8 1516 61

View Full Product Details