Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC46A3
Entrez Gene ID
283537
Gene Name
solute carrier family 46 member 3
Gene Aliases
FKSG16
Chromosome Location
Chr.13: 28700080 - 28719013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135919.1 NP_001129391.1 3 - 4 1606 81
NM_181785.3 NP_861450.1 3 - 4 1606 81
XM_005266361.2 XP_005266418.1 3 - 4 1284 81
GenBank mRNA AF315594.1 - 3 - 4 1190 81
AK093932.1 - 2 - 3 2034 81
AK098426.1 - 1 - 2 636 81
AK124604.1 - 1 - 2 82
BC036662.1 - 3 - 4 1568 81
BC060850.1 - 3 - 4 1427 81
BC068556.1 - 3 - 4 1434 81
BX537565.1 - 3 - 4 1417 81
CB851220.1 - 1 - 2 133 81

View Full Product Details