The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319043.1 | NP_001305972.1 | 1 - 2 | 152 | 82 |
NM_001319044.1 | NP_001305973.1 | 1 - 2 | 152 | 82 | |
NM_001319045.1 | NP_001305974.1 | 1 - 2 | 152 | 82 | |
NM_001319046.1 | NP_001305975.1 | 1 - 2 | 152 | 82 | |
NM_032025.4 | NP_114414.2 | 1 - 2 | 152 | 82 | |
XM_011513224.1 | XP_011511526.1 | 1 - 2 | 135 | 82 | |
GenBank mRNA | AF212241.1 | - | 1 - 2 | 101 | 82 |
AK298586.1 | - | 1 - 2 | 63 | 82 | |
DA382303.1 | - | 1 - 2 | 153 | 82 |