Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGHMBP2
Entrez Gene ID
3508
Gene Name
immunoglobulin mu binding protein 2
Gene Aliases
CATF1, CMT2S, HCSA, HMN6, SMARD1, SMUBP2, ZFAND7
Chromosome Location
Chr.11: 68903851 - 68940602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002180.2 NP_002171.2 7 - 8 1166 66
XM_005273974.2 XP_005274031.1 7 - 8 1130 66
XM_005273976.1 XP_005274033.1 7 - 8 1166 66
XM_017017669.1 XP_016873158.1 6 - 7 1022 66
XM_017017670.1 XP_016873159.1 3 - 4 927 66
XM_017017671.1 XP_016873160.1 7 - 8 1166 66
GenBank mRNA AB208802.1 - - 1108 66
AK312565.1 - 7 - 8 1124 66
BC000290.2 - 7 - 8 1079 66
BC025299.1 - 7 - 8 1111 66
BC080519.1 - 7 - 8 1111 66
BC105088.1 - 7 - 8 1103 66
BC105090.1 - 7 - 8 1103 66
L14754.1 - 7 - 8 1113 66

View Full Product Details