Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD10
Entrez Gene ID
11235
Gene Name
programmed cell death 10
Gene Aliases
CCM3, TFAR15
Chromosome Location
Chr.3: 167683891 - 167735690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007217.3 NP_009148.2 4 - 5 545 126
NM_145859.1 NP_665858.1 4 - 5 394 126
NM_145860.1 NP_665859.1 3 - 4 300 126
XM_005247086.4 XP_005247143.1 4 - 5 1280 126
XM_005247087.4 XP_005247144.1 4 - 5 1285 126
XM_005247088.3 XP_005247145.1 3 - 4 376 126
XM_006713485.3 XP_006713548.1 4 - 5 440 126
XM_011512368.2 XP_011510670.1 5 - 6 1353 126
XM_011512369.2 XP_011510671.1 5 - 6 1358 126
XM_017005644.1 XP_016861133.1 4 - 5 435 126
GenBank mRNA AF022385.1 - 3 - 4 300 126
AK291130.1 - 3 - 4 331 126
BC002506.1 - 4 - 5 557 126
BC016353.1 - 4 - 5 398 126
CR457107.1 - 2 - 3 147 126

View Full Product Details