The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253891.1 | NP_001240820.1 | 5 - 6 | 937 | 66 |
NM_032564.4 | NP_115953.2 | 6 - 7 | 1066 | 66 | |
XM_011545304.2 | XP_011543606.1 | 6 - 7 | 770 | 66 | |
GenBank mRNA | AB048286.2 | - | 6 - 7 | 1037 | 66 |
AB073384.1 | - | 5 - 6 | 614 | 66 | |
AF384161.2 | - | 6 - 7 | 814 | 66 | |
AK074203.1 | - | 2 - 3 | 331 | 66 | |
AK091870.1 | - | 1 - 2 | 2194 | 66 | |
AL834287.1 | - | 6 - 7 | 945 | 66 | |
AY358532.1 | - | 6 - 7 | 991 | 66 | |
AY780647.1 | - | 5 - 6 | 669 | 66 | |
BC015234.1 | - | 6 - 7 | 1055 | 66 | |
CR749377.1 | - | 2 - 3 | 4550 | 66 |