The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005651.3 | NP_005642.1 | 8 - 9 | 909 | 124 |
GenBank mRNA | AK289418.1 | - | 8 - 9 | 906 | 124 |
BC005355.1 | - | 8 - 9 | 906 | 124 | |
BX647341.1 | - | 5 - 6 | 2352 | 124 | |
U32989.1 | - | 8 - 9 | 907 | 124 |