The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164501.1 | NP_001157973.1 | 10 - 11 | 1710 | 101 |
NM_019843.3 | NP_062817.2 | 10 - 11 | 1734 | 101 | |
XM_005261686.2 | XP_005261743.1 | 10 - 11 | 1734 | 101 | |
XM_005261687.2 | XP_005261744.1 | 10 - 11 | 1715 | 101 | |
XM_005261688.2 | XP_005261745.1 | 10 - 11 | 1585 | 101 | |
XM_006724281.3 | XP_006724344.3 | 10 - 11 | 2011 | 101 | |
XM_011530280.1 | XP_011528582.1 | 10 - 11 | 1585 | 101 | |
XM_011530281.2 | XP_011528583.1 | 10 - 11 | 1806 | 101 | |
XM_017028863.1 | XP_016884352.1 | 10 - 11 | 2011 | 101 | |
XM_017028864.1 | XP_016884353.1 | 10 - 11 | 1806 | 101 | |
GenBank mRNA | AF240775.1 | - | 10 - 11 | 1564 | 101 |
AK025254.1 | - | 3 - 4 | 462 | 101 | |
AK314636.1 | - | 10 - 11 | 1735 | 101 | |
BC033028.1 | - | 10 - 11 | 1684 | 101 | |
CR456386.1 | - | 9 - 10 | 1543 | 101 |