The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164501.1 | NP_001157973.1 | 5 - 6 | 780 | 69 |
NM_019843.3 | NP_062817.2 | 5 - 6 | 804 | 69 | |
XM_005261686.2 | XP_005261743.1 | 5 - 6 | 804 | 69 | |
XM_005261687.2 | XP_005261744.1 | 5 - 6 | 785 | 69 | |
XM_005261688.2 | XP_005261745.1 | 5 - 6 | 655 | 69 | |
XM_006724281.3 | XP_006724344.3 | 5 - 6 | 1081 | 69 | |
XM_011530280.1 | XP_011528582.1 | 5 - 6 | 655 | 69 | |
XM_011530281.2 | XP_011528583.1 | 5 - 6 | 876 | 69 | |
XM_017028863.1 | XP_016884352.1 | 5 - 6 | 1081 | 69 | |
XM_017028864.1 | XP_016884353.1 | 5 - 6 | 876 | 69 | |
GenBank mRNA | AF240775.1 | - | 5 - 6 | 634 | 69 |
AK314636.1 | - | 5 - 6 | 805 | 69 | |
BC012734.2 | - | 5 - 6 | 657 | 69 | |
BC033028.1 | - | 5 - 6 | 754 | 69 | |
CR456386.1 | - | 4 - 5 | 613 | 69 |